Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formyltetrathiafulvalene 98.0+%, TCI America™
SDP

Catalog No. F02851G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F02851G Supplier TCI America Supplier No. F02851G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68128-94-9
Molecular Formula C7H4OS4
Molecular Weight (g/mol) 232.348
InChI Key NDJHQJARYXJFDI-UHFFFAOYSA-N
Synonym Formyl-TTF, [2,2′C-Bi(1,3-dithiolylidene)]-4-carboxaldehyde
PubChem CID 339970
IUPAC Name 2-(1,3-dithiol-2-ylidene)-1,3-dithiole-4-carbaldehyde
SMILES C1=CSC(=C2SC=C(S2)C=O)S1

Specifications

Melting Point 113°C
Color Black-Yellow
Quantity 1 g
Formula Weight 232.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Formyltetrathiafulvalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.