Learn More
Medchemexpress LLC HY-16209 1mg , Forodesine (hydrochloride) CAS:284490-13-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY162091MG
Encompass_Preferred
HY-16209 1mg Forodesine (hydrochloride) CAS: 284490-13-7 Forodesine hydrochloride (BCX-1777 hydrochloride) is a highly potent and orally active purine nucleoside phosphorylase (PNP) inhibitor with IC50 values ranging from 0.48 to 1.57 nM for human, mouse, rat, monkey and dog PNP. Forodesine hydrochloride is a potent human lymphocyte proliferation inhibitor. Forodesine hydrochloride could induce apoptosis in leukemic cells by increasing the dGTP levels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0968