Learn More
Cayman Chemical ForodesInhydrochlorIde 10mg
SDP
Catalog No. 502918430
Encompass_Preferred
A nucleoside derivative and PNP inhibitor (IC50s = 1.19, 0.48, 1.24, 0.66, and 1.57 nM for human, mouse, rat, monkey, and dog PNP, respectively); inhibits phytohemagglutinin A-, mixed lymphocyte reaction-, or IL-2-induced proliferation of isolated human PBLs in the presence of 2'-deoxyguanosine; inhibits the proliferation of CEM-SS T-ALL; prolongs survival in the hu-PBL-SCID mouse model of xenogeneic GVHD
Catalog No. 50-291-8430 Supplier Cayman Chemical Supplier No. 3960910 MG
A nucleoside derivative and PNP inhibitor (IC50s = 1.19, 0.48, 1.24, 0.66, and 1.57 nM for human, mouse, rat, monkey, and dog PNP, respectively); inhibits phytohemagglutinin A-, mixed lymphocyte reaction-, or IL-2-induced proliferation of isolated human PBLs in the presence of 2'-deoxyguanosine; inhibits the proliferation of CEM-SS T-ALL; prolongs survival in the hu-PBL-SCID mouse model of xenogeneic GVHD