Learn More
Medchemexpress LLC HY-128694 5mg Medchemexpress, Foropafant CAS:136468-36-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1286945MG
Encompass_Preferred
Medchemexpress, HY-128694 5mg Foropafant CAS:136468-36-5 Foropafant (SR27417) highly potent, competitive, selective and orally active antagonist of platelet-activating factor (PAF) receptor, with a Ki value of 57 pM for [3H]PAF binding, at least 5-fold lower than that of unlabeled PAF itself. Foropafant potently inhibits PAF-induced aggregation of rabbit and human platelets. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9757