Learn More
AccuStandard, Inc FORPIFICATION SOL 5X1ML
SDP
Catalog No. NC9470185
Encompass
Fortification Solution. 100 mL. 10 ?g/mL in 2-5% Nitric Acid.
Catalog No. NC9470185 Supplier AccuStandard, Inc Supplier No. M524FSPAK
Fortification Solution. 100 mL. 10 ?g/mL in 2-5% Nitric Acid.