Learn More
AdipoGen Forskolin, 66428-89-5, MFCD00082317, 1 mg
SDP
Catalog No. 50596447
Encompass_Preferred
MF: C22H34O7, MW: 410.5, ≥99%, Appearance: White to off-white solid. Potent, cell permeable adenylyl cyclase activator. Increases intracellular cAMP levels. Widely used tool to investigate cAMP as a second messenger. Inotropic and antihypertensive. Smooth muscle relaxant/vasodilator.
Catalog No. 50-596-447 Supplier AdipoGen Supplier No. AGCN20089M001
MF: C22H34O7, MW: 410.5, ≥99%, Appearance: White to off-white solid. Potent, cell permeable adenylyl cyclase activator. Increases intracellular cAMP levels. Widely used tool to investigate cAMP as a second messenger. Inotropic and antihypertensive. Smooth muscle relaxant/vasodilator.