Learn More
FUJIFILM Biosciences Forskolin, 10 mg, CAS # : 66575-29-9 Molecular Formula : C22H34O7 Molecular Weight : 410.50
Catalog No. NC3727633
Encompass_Preferred
>A kind of diterpenes isolated from Coleus forskohlii. It accelerates the activity of adenylate cyclase (rat liver membranes). In vivo, it accelerates cAMP secretion and exhibits hypotensive activity and bronchodilation.
Catalog No. NC3727633 Supplier FUJIFILM Biosciences Supplier No. 06702191
>A kind of diterpenes isolated from Coleus forskohlii. It accelerates the activity of adenylate cyclase (rat liver membranes). In vivo, it accelerates cAMP secretion and exhibits hypotensive activity and bronchodilation.