Learn More
Enzo Life Sciences Forskolin (100mg). CAS: 66575-29-9
SDP
Supplier:  Enzo Life Sciences BMLCN1000100
Encompass_Preferred
Activates adenylate cyclase by directly interacting with the catalytic unit of the enzyme leading to an increase in the intracellular concentration of cAMP. Widely used tool for the investigation of the role of cAMP as a second messenger with a broad range of potential therapeutic applications. Inotropic agent and vasodilator. Dose-dependent reduction of platelet aggregation. Inhibits ion channels by a mechanism that does not involve cAMP. Non-competitive inhibitor of nicotinic acetylcholine receptors. Induces neuronal differentiation in stem cells and several neuroblastoma. Alternative name: Colforsin. Purity: ≥97% (HPLC, TLC). Solubility: Soluble in ethyl acetate (10mg/ml), anhydrous DMSO (5mg/ml) or 100% ethanol (6mg/ml). DMSO is the recommended solvent for activation of cAMP. Long Term Storage: -20°C.
Catalog No. 50-201-0436