Learn More
Medchemexpress LLC HY-15371 100mg , Forskolin CAS:66575-29-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15371100MG
Encompass_Preferred
HY-15371 100mg Forskolin CAS: 66575-29-9 Forskolin (Coleonol) is a potent adenylate cyclase activator with an IC50 of 41 nM and an EC50 of 0.5 μM for type I adenylyl cyclase. Forskolin is also an inducer of intracellular cAMP formation. Forskolin induces differentiation of various cell types and activates pregnane X receptor (PXR) and FXR. Forskolin exerts a inotropic effect on the heart, and has platelet antiaggregatory and antihypertensive actions. Forskolin also induces autophagy[4][5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0157