Learn More
Cayman Chemical Forskolin, 66575-29-9, 10 mg
SDP
Supplier:  Cayman Chemical 1101810
Encompass_Preferred
Molecular Formula C22H34O7, Purity ≥98%, Formula Weight 410.5, Synonyms: Coleonol, HL 362, L 75-1362B, NSC 357088, NSC 375489; A naturally occurring diterpene that directly activates adenylyl cyclase to raise levels of cAMP in a wide variety of cell types; binds to type 1 adenylyl cyclase membranes with an IC50 value of 41 nM and demonstrates an EC50 value of 0.5 M in an activation assay.
Catalog No. NC1210583