Learn More
Enzo Life Sciences Forskolin, (10mg), CAS Number: 66575-29-9
SDP
Supplier:  Enzo Life Sciences BMLCN1000010
Encompass_Preferred
Adenylate cyclase activator. Alternative Name: Colforsin. Formula: C22H34O7. MW: 410.5. CAS Number: 66575-29-9. MI: 14: 2476. Purity: ≥97% (HPLC, TLC). Appearance: White solid. Solubility: Soluble in ethyl acetate (10mg/ml), anhydrous DMSO (5mg/ml) or 100% ethanol (6mg/ml). DMSO is the recommended solvent for activation of cAMP. Use/Stability: Stock solutions in DMSO are stable for up to 4 months when stored at +4°C. Do not dissolve in ethanol, which inhibits forskolin activation of adenylate cyclase. Long Term Storage: -20°C
Catalog No. NC9768491