Learn More
Apexbio Technology LLC Forskolin 66575-29-9 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B14215.1
Encompass_Preferred
Forskolin (CAS 66575-29-9) is a diterpenoid isolated from the plant Coleus forskohlii known to directly activate adenylate cyclase type I thereby elevating cellular cyclic AMP (cAMP) concentrations It exhibits an IC50 of approximately 41 nM against adenylate cyclase By increasing intracellular cAMP forskolin modulates signaling pathways involved in inflammation and oxidative stress reducing macrophage activation and subsequent production of thromboxane B2 and superoxide Forskolin has been explored in various experimental models and clinical contexts including cardiovascular disorders diabetes mellitus and asthma research
Catalog No. 50-000-61520