Learn More
AdipoGen Forskolin
SDP
Supplier:  AdipoGen AGCN20089M005
Encompass_Preferred
Chemical. CAS 66428-89-5 and 66575-29-9. Formula C22H34O7. MW 410.5. Isolated from Coleus forskohlii. Potent, cell permeable adenylyl cyclase activator. Increases intracellular cAMP levels. Widely used tool to investigate cAMP as a second messenger. Inotropic and antihypertensive. Smooth muscle relaxant/vasodilator. Glucose transporter inhibitor. Platelet aggregation inhibitor. Stimulates lipolysis in fat cells. Non-competitive nicotinic acetylcholine receptors inhibitor. MAP kinase inhibitor. Upregulates mitochondrial uncoupling protein UCP mRNA levels in brown adipose tissue. Autophagy inhibitor. Hedgehog signaling inhibitor. Has antiglaucoma potential. Promotes neuronal differentiation of NSCs.
Catalog No. 50-256-2664