Learn More
Abcam Forskolin, Adenylyl cyclase activator, 5MG
Catalog No. NC1456149
Encompass_Preferred
MW 410.5 Da. Cell-permeable activator of adenylyl cyclase. Interacts directly with the catalytic subunit of the enzyme to increase intracellular cAMP levels. Positive inotropic, platelet antiaggregatory and antihypertensive actions in vivo. 3D Growth matrix component used in liver organoid generation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1456149 Supplier Abcam Supplier No. AB1200585MG
MW 410.5 Da. Cell-permeable activator of adenylyl cyclase. Interacts directly with the catalytic subunit of the enzyme to increase intracellular cAMP levels. Positive inotropic, platelet antiaggregatory and antihypertensive actions in vivo. 3D Growth matrix component used in liver organoid generation.
The product is subject to the following: Abcam Restricted Use Statement