Learn More
Abcam Forskolin, Adenylyl cyclase activator, 50MG
Catalog No. 502584714
Encompass_Preferred
MW 410.5 Da. Cell-permeable activator of adenylyl cyclase. Interacts directly with the catalytic subunit of the enzyme to increase intracellular cAMP levels. Positive inotropic, platelet antiaggregatory and antihypertensive actions in vivo. 3D Growth matrix component used in liver organoid generation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4714 Supplier Abcam Supplier No. AB12005850MG
MW 410.5 Da. Cell-permeable activator of adenylyl cyclase. Interacts directly with the catalytic subunit of the enzyme to increase intracellular cAMP levels. Positive inotropic, platelet antiaggregatory and antihypertensive actions in vivo. 3D Growth matrix component used in liver organoid generation.
The product is subject to the following: Abcam Restricted Use Statement