Learn More
Medchemexpress LLC HY-N0029 10mg Medchemexpress, Forsythoside B CAS:81525-13-5 Purity:>98%
SDP
Catalog No. 501933360
Encompass_Preferred
Medchemexpress, HY-N0029 10mg Forsythoside B CAS:81525-13-5 Forsythoside B is a phenylethanoid glycoside isolated from the leaves of Lamiophlomis rotata Kudo, a Chinese folk medicinal plant for treating inflammatory diseases and promoting blood circulation. Forsythoside B could inhibit TNF-alpha , IL-6 , IκB and modulate NF-κB . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3360 Supplier Medchemexpress LLC Supplier No. HYN002910MG
Medchemexpress, HY-N0029 10mg Forsythoside B CAS:81525-13-5 Forsythoside B is a phenylethanoid glycoside isolated from the leaves of Lamiophlomis rotata Kudo, a Chinese folk medicinal plant for treating inflammatory diseases and promoting blood circulation. Forsythoside B could inhibit TNF-alpha , IL-6 , IκB and modulate NF-κB . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.