Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Forsythoside E | 93675-88-8 | 5 MG
SDP

Catalog No. 5000418779
Encompass_Preferred

Forsythoside E is a phenylethanoid glycoside isolated from the fruits of Forsythia suspensa (Thunb.) Vahl. This product is intended for research use only.

  • Phenylethanoid glycoside
  • Isolated from Forsythia suspensa (Thunb.) Vahl.
  • Purity of 98.57%
  • Appearance: solid, white to off-white
  • Recommended storage: 4°C, protect from light
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (protect from light)
  • Soluble in DMSO

Catalog No. 50-004-18779 Supplier Medchemexpress LLC Supplier No. HYN21735MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Forsythoside E is a phenylethanoid glycoside isolated from the fruits of Forsythia suspensa (Thunb.) Vahl. This product is intended for research use only.

  • Phenylethanoid glycoside
  • Isolated from Forsythia suspensa (Thunb.) Vahl.
  • Purity of 98.57%
  • Appearance: solid, white to off-white
  • Recommended storage: 4°C, protect from light
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (protect from light)
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.