Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Forsythoside I | 1177581-50-8 | 99.9% | 624.59 | C29H36O15 | 10 MG
SDP

Catalog No. 5000289588
Encompass_Preferred

Forsythoside I is a caffeoyl phenylethanoid glycoside used in biological research for its reported anti-inflammatory activity, including inhibition of pro-inflammatory cytokines and modulation of the TXNIP/NLRP3 inflammasome pathway.

  • Used as an anti-inflammatory research compound that modulates TXNIP/NLRP3 inflammasome.
  • Caffeoyl phenylethanoid glycoside; CAS number 1177581-50-8.
  • Molecular formula C29H36O15 and molecular weight 624.59 g·mol⁻¹.
  • High purity (99.94%) as supplied by the manufacturer.
  • White to off-white solid supplied as 10 mg in a sealed vial.
  • Soluble in DMSO (100 mg/mL); in vivo formulations reported include 10% DMSO with PEG300 or SBE-β-CD.
  • Store solid at 4°C and protect from light; store solutions at -80°C for long term (up to 6 months) or -20°C for short term (up to 1 month).

Catalog No. 50-002-89588 Supplier Medchemexpress LLC Supplier No. HYN504210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Forsythoside I is a caffeoyl phenylethanoid glycoside used in biological research for its reported anti-inflammatory activity, including inhibition of pro-inflammatory cytokines and modulation of the TXNIP/NLRP3 inflammasome pathway.

  • Used as an anti-inflammatory research compound that modulates TXNIP/NLRP3 inflammasome.
  • Caffeoyl phenylethanoid glycoside; CAS number 1177581-50-8.
  • Molecular formula C29H36O15 and molecular weight 624.59 g·mol⁻¹.
  • High purity (99.94%) as supplied by the manufacturer.
  • White to off-white solid supplied as 10 mg in a sealed vial.
  • Soluble in DMSO (100 mg/mL); in vivo formulations reported include 10% DMSO with PEG300 or SBE-β-CD.
  • Store solid at 4°C and protect from light; store solutions at -80°C for long term (up to 6 months) or -20°C for short term (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.