Learn More
Inorganic Ventures 300.0 LAB FORTIF STD 125mL
SDP
Catalog No. 50723008
Encompass_Preferred
Matrix: Water (H2O), Dilution: 1:100 to 1:1000, Analyte: Br- 1000 ug/mL, F- 200 ug/mL, SO4-2 1500 ug/mL, Phosphate as P 500 ug/mL, Nitrite as N, Nitrate as N, and Cl- 300 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-008 Supplier Inorganic Ventures Supplier No. 300LFSA125ML
Matrix: Water (H2O), Dilution: 1:100 to 1:1000, Analyte: Br- 1000 ug/mL, F- 200 ug/mL, SO4-2 1500 ug/mL, Phosphate as P 500 ug/mL, Nitrite as N, Nitrate as N, and Cl- 300 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search