Learn More
Inorganic Ventures 200.7WW LFS SOLUTION 1 125mL
SDP
Catalog No. 50723864
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Dilution: 1:100, Analyte: Al, Mg, Se, Tl, and Ce 200 ug/mL each, Ba, Zn, Be, Cd, Co, Li, Mn, and Sr 20 ug/mL each, B, Cu, and V 30 ug/mL, Ca, and Pb 100 ug/mL each, Cr 40 ug/mL each, Hg 70 ug/mL, P 600 ug/mL, Na, and Fe 300 ug/mL, As 80 ug/mL, Ni 50 ug/mL, K 1000 ug/mL, and Ag 7.5 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-864 Supplier Inorganic Ventures Supplier No. WWLFS1125ML
Matrix: Nitric Acid (HNO3), Dilution: 1:100, Analyte: Al, Mg, Se, Tl, and Ce 200 ug/mL each, Ba, Zn, Be, Cd, Co, Li, Mn, and Sr 20 ug/mL each, B, Cu, and V 30 ug/mL, Ca, and Pb 100 ug/mL each, Cr 40 ug/mL each, Hg 70 ug/mL, P 600 ug/mL, Na, and Fe 300 ug/mL, As 80 ug/mL, Ni 50 ug/mL, K 1000 ug/mL, and Ag 7.5 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search