Learn More
Inorganic Ventures 200.7WW LFS SOLUTION 2 125mL
SDP
Catalog No. 50723865
Encompass_Preferred
Matrix: Nitric Acid / Hydrofluoric Acid) (HNO3 / HF), Dilution: 1:100, Analyte: Sb 80 ug/mL, SiO2 200 ug/mL, Ti 20 ug/mL, Mo 40 ug/mL, and Sn 70 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-865 Supplier Inorganic Ventures Supplier No. WWLFS2125ML
Matrix: Nitric Acid / Hydrofluoric Acid) (HNO3 / HF), Dilution: 1:100, Analyte: Sb 80 ug/mL, SiO2 200 ug/mL, Ti 20 ug/mL, Mo 40 ug/mL, and Sn 70 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search