Learn More
Med Vet International Purina Fortiflora Caninie Probiotic Supplement, 30 Packets, 6/Box
Catalog No. 502828248
Encompass_Preferred
Purina Veterinary Diets FortiFlora has been formulated to achieve the following characteristics: Contains a guaranteed level of viable microorganisms Proprietary microencapsulation process for enhanced stability Proven to promote normal intestinal microflora Promotes a strong immune system Shown to be safe for use in dogs Contains high levels of antioxidant Vitamins A, E, and C Excellent palatability Medical Indications FortiFlora is recommended as a nutritional supplement to help with the following: Diarrhea associated with microflora imbalance Acute enteritis Diarrhea associated with stress, antibiotic therapy, and diet change Poor fecal quality in puppies To promote a strong immune system Medical Contraindications Dogs with food allergies • Severely immune-compromised dogs
Catalog No. 50-282-8248 Supplier Med Vet International Supplier No. PHVFORTICAN
Purina Veterinary Diets FortiFlora has been formulated to achieve the following characteristics: Contains a guaranteed level of viable microorganisms Proprietary microencapsulation process for enhanced stability Proven to promote normal intestinal microflora Promotes a strong immune system Shown to be safe for use in dogs Contains high levels of antioxidant Vitamins A, E, and C Excellent palatability Medical Indications FortiFlora is recommended as a nutritional supplement to help with the following: Diarrhea associated with microflora imbalance Acute enteritis Diarrhea associated with stress, antibiotic therapy, and diet change Poor fecal quality in puppies To promote a strong immune system Medical Contraindications Dogs with food allergies • Severely immune-compromised dogs