Learn More
Electron Microscopy Sciences Fortress™ Sample Loader for Low Profile
SDP
Catalog No. 5032093
Encompass
Securely holds the Fortress high Profile holder under a stereo microscope. Viewing from 2 angles without changing focus on the microscope.
Catalog No. 50-320-93 Supplier Electron Microscopy Sciences Supplier No. 7595750
Securely holds the Fortress high Profile holder under a stereo microscope. Viewing from 2 angles without changing focus on the microscope.