Learn More
Electron Microscopy Sciences Fortress™ Sample Loader for Low Profile
SDP
Supplier:  Electron Microscopy Sciences 7595650
Encompass
Securely holds the Fortress low Profile holder under a stereo microscope. Viewing from 2 angles without changing focus on the microscope.
Catalog No. 50-320-95