Learn More
Sigma Aldrich Fine Chemicals Biosciences Glass Sterile Fortuna Optima Syringes with Interchangeable Barrels and Luer Lock Tips | 1 ML, 20 ML, 50 ML Capacities | Clear Color
Catalog No. NC2584864
Encompass
The glass Fortuna Optima syringes are sterile and can accurately transfer volumes of fluids. They are available with 1 mL, 20 mL, and 50 mL capacities. The interchangeable glass barrels and plungers allow customization. The syringes have luer lock tips for secure connections. The colored rings on the plungers enable easy volume measurement. The flat flanges on the glass barrels prevent rolling when placed on flat surfaces.
Catalog No. NC2584864 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3145601EA
The glass Fortuna Optima syringes are sterile and can accurately transfer volumes of fluids. They are available with 1 mL, 20 mL, and 50 mL capacities. The interchangeable glass barrels and plungers allow customization. The syringes have luer lock tips for secure connections. The colored rings on the plungers enable easy volume measurement. The flat flanges on the glass barrels prevent rolling when placed on flat surfaces.