Learn More
Sigma Aldrich Fine Chemicals Biosciences fORTUNAR OptifixR Solvent1EA
Catalog No. 502906070
Encompass
for dispensing solvents featuresDispensing piston made of glassVisible dispensing process by the use of a dispensing cylinder made of glass coated with plastic-protection against breakagefIX adapter made of white PP up to 10 mL fIX adapter made of solid white PP from 30 mL sizeValve block made of PTfE no metal springs in the valve - no corrosion Ergonomic finger protection capfast and easy volume setting with additional fine adjustmentOptifix Solvent dispenser available up to 500 mLSupplied as Optifix Solvent dispenser 2 PP adapters GL40 GL45 1 PTfE intake tube
Catalog No. 50-290-6070 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2601261EA
for dispensing solvents featuresDispensing piston made of glassVisible dispensing process by the use of a dispensing cylinder made of glass coated with plastic-protection against breakagefIX adapter made of white PP up to 10 mL fIX adapter made of solid white PP from 30 mL sizeValve block made of PTfE no metal springs in the valve - no corrosion Ergonomic finger protection capfast and easy volume setting with additional fine adjustmentOptifix Solvent dispenser available up to 500 mLSupplied as Optifix Solvent dispenser 2 PP adapters GL40 GL45 1 PTfE intake tube