Learn More
Sigma Aldrich Fine Chemicals Biosciences fORTUNAR Optima glass syri
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z31455210EA
Encompass
Interchangeable barrels and plungersflanges flat on two sides to prevent syringe rollingColored ring on plunger ends for easy volume measurementConformity certified by H markGlass Luer - standard Luer hubLuer lock - metal Luer lock tip on glass syringe easy twist locks Luer needle onto syringe
Catalog No. 50-290-0003