Learn More
Sigma Aldrich Fine Chemicals Biosciences fORTUNAR Optima glass syri
Catalog No. 502901361
Encompass
Interchangeable barrels and plungersflanges flat on two sides to prevent syringe rollingColored ring on plunger ends for easy volume measurementConformity certified by H markGlass Luer - standard Luer hubLuer lock - metal Luer lock tip on glass syringe easy twist locks Luer needle onto syringe
Catalog No. 50-290-1361 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z31430710EA
Interchangeable barrels and plungersflanges flat on two sides to prevent syringe rollingColored ring on plunger ends for easy volume measurementConformity certified by H markGlass Luer - standard Luer hubLuer lock - metal Luer lock tip on glass syringe easy twist locks Luer needle onto syringe