Learn More
Pyramex™ Forum™ Safety Eyewear
Catalog No. 19155798 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Clear
Indoor/Outdoor, Mirror
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-798 Clear
19-155-802 Indoor/Outdoor, Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-155-798 Supplier Pyramex™ Supplier No. SB6610D
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Fashionable stream line design incorporates the elements of safety in trend-setting eyewear
  • Soft PVC nose piece generates a cushion for a comfort fit
  • Metal emblem on temple presents this premium eyewear at the top of its class
  • 9.5 base curve lens provides excellent side protection
  • Scratch-resistant polycarbonate provides 99.9% UV protection
  • Passes MIL-PRF 32432 High Velocity Impact Standards
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Certifications/Compliance (Europe) EN-166
    Lens Tint Clear
    Frame Color Black
    Weight (Metric) 26 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate