Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Forward Vision Mobile Table
GREENER_CHOICESCI_ED

Catalog No. S80774 Shop All Diversified Spaces Products
Change view
Click to view available options
Height (English):
36 in.
Material:
Solid Maple
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Material
S80774 36 in. Solid Maple
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S80774 Supplier Diversified Spaces™ Supplier No. FVT74M36C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed by taking our most popular work station and making it into a mobile table. The Forward Vision Mobile Table provides flexibility in the classroom by reconfiguring your space.

  • Legs and aprons are finished with chemical resistant, earth-friendly UV finish
  • Aprons feature heavy-duty, 13 gauge steel corner braces
  • 2.25 in. square legs with 5/16 in. hanger bolts are attached to apron with nuts and washers
  • Industrial locking casters
  • Top Surface: 3/4 in. Phenolic

Specifications

Product Type Mobile Classroom Workstation
Color Maple
Finish Phenolic Top
With Casters Yes
Footrest No
With Tilt No
Surface Coating Phenolic
Certifications/Compliance MAS Green
Model Forward Vision Table with Phenolic top
Warranty Limited Lifetime
For Use With (Application) Classroom Flexibility by Reconfiguring The Space.
Height (English) 36 in.
Thickness (English) 3/4 in.
Depth (English) 44 in.
Weight (English) 213 lb.
Material Solid Maple
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.