Learn More
Aviva Systems Biology Corp FOS Antibody - C-terminal region (P100639_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100639_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against.S. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FOS. Alternate names: p55, AP-1, C-FOSApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat, Sheep
Catalog No. 50-014-62675