Learn More
Cell Signaling Technology Fos Family Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. 502051232
Encompass
The Fos Family Antibody Sampler Kit provides an economical means to evaluate the Fos family of transcription factors. These proteins, including c-Fos, FosB, Fos-related antigen 1 (FRA1), and Fos-related antigen 2 (FRA2), dimerize with Jun proteins to form the AP-1 transcription factor, which plays a critical role in cellular growth, differentiation, and transformation.
  • Includes primary antibodies for the analysis of Fos, FosB, FRA1, Phospho-Fos (Ser32), and Phospho-FRA1 (Ser265)
  • Contains HRP-linked anti-rabbit IgG secondary antibody
  • Validated for multiple applications including western blotting, immunoprecipitation, immunohistochemistry, immunofluorescence, flow cytometry, and chromatin immunoprecipitation
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol and less than 0.02% sodium azide
Catalog No. 50-205-1232 Supplier Cell Signaling Technology Supplier No. 8333T
The Fos Family Antibody Sampler Kit provides an economical means to evaluate the Fos family of transcription factors. These proteins, including c-Fos, FosB, Fos-related antigen 1 (FRA1), and Fos-related antigen 2 (FRA2), dimerize with Jun proteins to form the AP-1 transcription factor, which plays a critical role in cellular growth, differentiation, and transformation.
  • Includes primary antibodies for the analysis of Fos, FosB, FRA1, Phospho-Fos (Ser32), and Phospho-FRA1 (Ser265)
  • Contains HRP-linked anti-rabbit IgG secondary antibody
  • Validated for multiple applications including western blotting, immunoprecipitation, immunohistochemistry, immunofluorescence, flow cytometry, and chromatin immunoprecipitation
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol and less than 0.02% sodium azide