Learn More
Laxco™ FOS150 Series Fiber Optic Light Source
SDP
Catalog No. p-7071650 Shop All Laxco Inc Products
Change view
Click to view available options
Type:
Bifurcated Fiber Optic Guides
Fiber Optic Ring Guide
Single Fiber Optic Light Guide
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
PIFOS150S Single Fiber Optic Light Guide
PIFOS150IB Bifurcated Fiber Optic Guides
PIFOS150R Fiber Optic Ring Guide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PIFOS150S Supplier Laxco™ Supplier No. PIFOS150S
Includes
Power cord, Spare bulb, Allen key, Light guide, Light source
Laxco™ FOS Series is 150w fiber optic light source with exchangeable guides for industrial, manufacturing and dissection applications.
  • Infrared filter placed directly in front of bulb reduces IR light rays that produce heat at sample
  • Adjustable focusing lens focuses 150w of illumination to a fine point, or broaden it to be a flood for (single and bifurcated options only)
  • Fan cooled and thermal fuse protected
  • Heat shield, hinged lid allows for easy access to replacing bulb and protection from bulb heat
  • Large neural knob securely locks light guides in place
Certifications
Quality Proformance Certification
Warranty and Services
2 Year - Electronic, Mechanical, Optical with option to extend to 5 years
Specifications
Type Single Fiber Optic Light Guide
Dimensions (L x W x H) 56.8 x 27.2 x 13 cm (22 x 10.7 x 5.11 in.)
For Use With (Application) Manufacturing, Research, Laboratory
Lighting Type 150 watt Halogen
Voltage 110 V, Internal
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase