Learn More
Medchemexpress LLC Fosamprenavir (calcium salt) | 226700-81-8 | MFCD08141843 | ≥98.0% | 623.67 g/mol | C25H34CaN3O9PS | 10 MG
SDP
Catalog No. 5000299206
Encompass_Preferred
Fosamprenavir calcium salt is the calcium salt form of fosamprenavir, an orally active prodrug of the HIV-1 protease inhibitor amprenavir. It is hydrolyzed by cellular phosphatases to release the active drug and is used as a research reagent for studies of HIV-1 protease inhibition and antiviral activity.
  • Orally active prodrug converted to amprenavir.
  • Inhibits HIV-1 protease, blocking viral maturation.
  • Calcium salt form suitable for research and analytical use.
  • Reported high purity available for analytical applications.
  • Supplied as a research-grade reagent with an associated safety data sheet.
Catalog No. 50-002-99206 Supplier Medchemexpress LLC Supplier No. HY1743110MG
Add to Cart
Add to Cart
Fosamprenavir calcium salt is the calcium salt form of fosamprenavir, an orally active prodrug of the HIV-1 protease inhibitor amprenavir. It is hydrolyzed by cellular phosphatases to release the active drug and is used as a research reagent for studies of HIV-1 protease inhibition and antiviral activity.
  • Orally active prodrug converted to amprenavir.
  • Inhibits HIV-1 protease, blocking viral maturation.
  • Calcium salt form suitable for research and analytical use.
  • Reported high purity available for analytical applications.
  • Supplied as a research-grade reagent with an associated safety data sheet.