Learn More
Apexbio Technology LLC Fosaprepitant(Synonyms: Fosaprepitant dimeglumine, Emend IV, L-758298, MK-0517, Fosaprepitant meglumine, Ivemend), 10mM (in 1mL DMSO), CAS: 172673-20-0.
SDP
Catalog No. 5000060407
Encompass_Preferred
Fosaprepitant (CAS 172673-20-0) also known as MK-0517 or L-758 298 is a selective neurokinin-1 (NK-1) receptor antagonist exerting its pharmacological effects primarily through its active metabolite aprepitant This antagonism blocks substance P-mediated biological effects including vomiting responses In established preclinical models such as the cisplatin-triggered emesis in ferrets fosaprepitant demonstrated activity in mitigating chemotherapy-induced vomiting Given aprepitant s favorable penetration into the central nervous system fosaprepitant exhibits high NK-1 receptor affinity and potentiates antiemetic actions of agents such as dexamethasone and granisetron It is used in combination therapies with 5-HT3 antagonists and corticosteroids for chemotherapy-induced nausea and vomiting (CINV)
Catalog No. 50-000-60407 Supplier Apexbio Technology LLC Supplier No. B12725.1
Fosaprepitant (CAS 172673-20-0) also known as MK-0517 or L-758 298 is a selective neurokinin-1 (NK-1) receptor antagonist exerting its pharmacological effects primarily through its active metabolite aprepitant This antagonism blocks substance P-mediated biological effects including vomiting responses In established preclinical models such as the cisplatin-triggered emesis in ferrets fosaprepitant demonstrated activity in mitigating chemotherapy-induced vomiting Given aprepitant s favorable penetration into the central nervous system fosaprepitant exhibits high NK-1 receptor affinity and potentiates antiemetic actions of agents such as dexamethasone and granisetron It is used in combination therapies with 5-HT3 antagonists and corticosteroids for chemotherapy-induced nausea and vomiting (CINV)