Learn More
Medchemexpress LLC Fosaprepitant dimeglumine | 265121-04-8 | 98.2% | C₃₇H₅₆F₇N₆O₁₆P | 100 MG
SDP
Catalog No. 5000355043
Encompass_Preferred
Fosaprepitant dimeglumine (MK-0517) is a proagent of Aprepitant and functions as a neurokinin-1 receptor antagonist. It is developed for the prevention of chemotherapy-induced nausea and vomiting (CINV). For research use only, this product is not sold to patients.
  • Proagent of Aprepitant.
  • Neurokinin-1 receptor antagonist.
  • Developed for preventing chemotherapy-induced nausea and vomiting (CINV).
  • Increases the antinociceptive effect in rats when administered intraperitoneally.
Catalog No. 50-003-55043 Supplier Medchemexpress LLC Supplier No. HY14407A100MG
Fosaprepitant dimeglumine (MK-0517) is a proagent of Aprepitant and functions as a neurokinin-1 receptor antagonist. It is developed for the prevention of chemotherapy-induced nausea and vomiting (CINV). For research use only, this product is not sold to patients.
  • Proagent of Aprepitant.
  • Neurokinin-1 receptor antagonist.
  • Developed for preventing chemotherapy-induced nausea and vomiting (CINV).
  • Increases the antinociceptive effect in rats when administered intraperitoneally.