Learn More
Aviva Systems Biology Corp FOSB Antibody - C-terminal region (ARP32519_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32519_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against.SB. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human FOSB. Alternate names: AP-1, G0S3, GOS3, GOSBApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-54631