Learn More
Medchemexpress LLC Fosbretabulin disodium | 168555-66-6 | 99.6% | 440.29 | 100MG
SDP
Catalog No. 5000386667
Encompass_Preferred
Fosbretabulin disodium is a tubulin destabilizing agent and a Combretastatin A4 proagent. It selectively targets endothelial cells to induce regression of nascent tumor neovessels, reduce tumor blood flow, and cause central tumor necrosis. This compound shows significant biological activity in vitro against various cancer cell lines and in vivo by increasing mean arterial blood pressure and reducing tumor blood flow.
  • Inhibits growth of multiple cancer cell lines (leukemia, pancreas, neuroblast, thyroid, lung-NSC, prostate, pharynx)
  • Increases mean arterial blood pressure
  • Reduces blood flow in tumors
Catalog No. 50-003-86667 Supplier Medchemexpress LLC Supplier No. HY17449100MG
Fosbretabulin disodium is a tubulin destabilizing agent and a Combretastatin A4 proagent. It selectively targets endothelial cells to induce regression of nascent tumor neovessels, reduce tumor blood flow, and cause central tumor necrosis. This compound shows significant biological activity in vitro against various cancer cell lines and in vivo by increasing mean arterial blood pressure and reducing tumor blood flow.
  • Inhibits growth of multiple cancer cell lines (leukemia, pancreas, neuroblast, thyroid, lung-NSC, prostate, pharynx)
  • Increases mean arterial blood pressure
  • Reduces blood flow in tumors