Learn More
Medchemexpress LLC Fosbretabulin disodium | 168555-66-6 | 99.9% | 440.29 | 25 MG
SDP
Catalog No. 5000384335
Encompass_Preferred
Fosbretabulin disodium is a tubulin destabilizing agent and a Combretastatin A4 proagent. It selectively targets endothelial cells, inducing the regression of nascent tumor neovessels, reducing tumor blood flow, and causing central tumor necrosis. This agent is intended for research use only.
  • Acts as a tubulin destabilizing agent
  • Functions as a Combretastatin A4 proagent
  • Selectively targets endothelial cells
  • Induces regression of nascent tumor neovessels
  • Reduces tumor blood flow
  • Causes central tumor necrosis
  • Inhibits growth of various cell lines in vitro
  • Increases mean arterial blood pressure and reduces heart rate in vivo
  • Reduces tumor blood flow in vivo
Catalog No. 50-003-84335 Supplier Medchemexpress LLC Supplier No. 25MGHY17449
Add to Cart
Add to Cart
Fosbretabulin disodium is a tubulin destabilizing agent and a Combretastatin A4 proagent. It selectively targets endothelial cells, inducing the regression of nascent tumor neovessels, reducing tumor blood flow, and causing central tumor necrosis. This agent is intended for research use only.
  • Acts as a tubulin destabilizing agent
  • Functions as a Combretastatin A4 proagent
  • Selectively targets endothelial cells
  • Induces regression of nascent tumor neovessels
  • Reduces tumor blood flow
  • Causes central tumor necrosis
  • Inhibits growth of various cell lines in vitro
  • Increases mean arterial blood pressure and reduces heart rate in vivo
  • Reduces tumor blood flow in vivo