Learn More
Medchemexpress LLC Fosbretabulin disodium | 168555-66-6 | MFCD00943911 | 99.9% | 440.29 g/mol | C18H19Na2O8P | 10 MG
SDP
Catalog No. 5000298710
Encompass_Preferred
Fosbretabulin disodium is a water-soluble phosphate prodrug of combretastatin A4 that destabilizes microtubules and acts as a vascular-disrupting agent used in preclinical oncology research. It selectively targets tumor endothelial cells, reduces tumor blood flow, and induces central tumor necrosis; supplied as a disodium phosphate salt for laboratory use.
  • Microtubule-destabilizing vascular-disruptor mechanism.
  • Water-soluble disodium phosphate prodrug form.
  • High purity (≈99.9%).
  • Molecular weight 440.29 g/mol; formula C18H19Na2O8P.
  • Available in small research quantities (1-100 mg) and 10 mM aqueous solutions.
  • Intended for preclinical and in vitro research applications.
Catalog No. 50-002-98710 Supplier Medchemexpress LLC Supplier No. HY1744910MG
Fosbretabulin disodium is a water-soluble phosphate prodrug of combretastatin A4 that destabilizes microtubules and acts as a vascular-disrupting agent used in preclinical oncology research. It selectively targets tumor endothelial cells, reduces tumor blood flow, and induces central tumor necrosis; supplied as a disodium phosphate salt for laboratory use.
  • Microtubule-destabilizing vascular-disruptor mechanism.
  • Water-soluble disodium phosphate prodrug form.
  • High purity (≈99.9%).
  • Molecular weight 440.29 g/mol; formula C18H19Na2O8P.
  • Available in small research quantities (1-100 mg) and 10 mM aqueous solutions.
  • Intended for preclinical and in vitro research applications.