Learn More
Apexbio Technology LLC Foscarnet Sodium, 1g. Cas: 63585-09-1 MFCD: MFCD00013973. Reverse transcriptase inhibitor
SDP
Supplier:  Apexbio Technology LLC B19461000
Encompass_Preferred
Foscarnet Sodium inhibits viral RNA polymerases, reverse transcriptase, and DNA polymerases through noncompetitive inhibition with dNTPs. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5291