Learn More
Selleck Chemical LLC Foscarnet Sodium 25mg 63585-09-1 Phosphonoformate
SDP
Catalog No. 501948177
Encompass_Preferred
Foscarnet Sodium (Phosphonoformate) inhibits viral RNA polymerases, reverse transcriptase, and DNA polymerases through noncompetitive inhibition with dNTPs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8177 Supplier Selleck Chemical LLC Supplier No. S307625MG
Foscarnet Sodium (Phosphonoformate) inhibits viral RNA polymerases, reverse transcriptase, and DNA polymerases through noncompetitive inhibition with dNTPs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.