Learn More
Selleck Chemical LLC Fosfomycin calcium S3710-25mg
SDP
Catalog No. 503110777
Encompass_Preferred
Fosfomycin (phosphomycin phosphonomycin) is bactericidal and inhibits bacterial cell wall biogenesis by inactivating the enzyme UDP-N-acetylglucosamine-3-enolpyruvyltransferase also known as MurA It enters the bacterial cell through the glycerophosphate transporter
Catalog No. 50-311-0777 Supplier Selleck Chemical LLC Supplier No. S371025MG
Fosfomycin (phosphomycin phosphonomycin) is bactericidal and inhibits bacterial cell wall biogenesis by inactivating the enzyme UDP-N-acetylglucosamine-3-enolpyruvyltransferase also known as MurA It enters the bacterial cell through the glycerophosphate transporter