Learn More
Apexbio Technology LLC Fosfomycin Disodium 26016-99-9 25mg
SDP
Catalog No. 5000092691
Encompass_Preferred
Fosfomycin Disodium (CAS 26016-99-9) is a small-molecule inhibitor targeting bacterial cell wall biosynthesis It is designed to irreversibly inhibit the initial enzymatic step in peptidoglycan synthesis disrupting bacterial cell structure Fosfomycin Disodium exerts its biological activity primarily through irreversible inhibition of cell wall synthesis In in vitro studies Fosfomycin Disodium demonstrates inhibitory activity against drug-resistant bacterial strains with reported IC50 values typically in the low micromolar range depending on bacterial species and experimental conditions Based on these pharmacological properties Fosfomycin Disodium holds research potential in studies of bacterial cell wall synthesis antimicrobial resistance pathways and central nervous system infection models
Catalog No. 50-000-92691 Supplier Apexbio Technology LLC Supplier No. C649725
Fosfomycin Disodium (CAS 26016-99-9) is a small-molecule inhibitor targeting bacterial cell wall biosynthesis It is designed to irreversibly inhibit the initial enzymatic step in peptidoglycan synthesis disrupting bacterial cell structure Fosfomycin Disodium exerts its biological activity primarily through irreversible inhibition of cell wall synthesis In in vitro studies Fosfomycin Disodium demonstrates inhibitory activity against drug-resistant bacterial strains with reported IC50 values typically in the low micromolar range depending on bacterial species and experimental conditions Based on these pharmacological properties Fosfomycin Disodium holds research potential in studies of bacterial cell wall synthesis antimicrobial resistance pathways and central nervous system infection models