Learn More
Selleck Chemical LLC Fosfomycin Disodium S5048-25mg
SDP
Catalog No. 503110162
Encompass_Preferred
Fosfomycin (Phosphonemycin) is a bactericidal low-molecular weight broad-spectrum antibiotic with putative activity against several bacteria including multidrug-resistant Gram-negative bacteria by irreversibly inhibiting an early stage in cell wall synthesis
Catalog No. 50-311-0162 Supplier Selleck Chemical LLC Supplier No. S504825MG
Fosfomycin (Phosphonemycin) is a bactericidal low-molecular weight broad-spectrum antibiotic with putative activity against several bacteria including multidrug-resistant Gram-negative bacteria by irreversibly inhibiting an early stage in cell wall synthesis