Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fosfomycin disodium, 90%

Catalog No. AAJ6660203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6660203 1 g
AAJ6660206 5 g
AAJ6660214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6660203 Supplier Thermo Scientific Chemicals Supplier No. J6660203
Add to Cart

missing translation for 'validMainframeMsg'

Prevents peptidoglycan synthesis by inhibiting MurA | CAS: 26016-99-9 | C3H5Na2O4P | 182.02 g/mol

Prevents peptidoglycan synthesis by inhibiting MurA, an enzyme responsible for synthesizing N-acetylmuramic acid or NAM, a major component of peptidoglycan

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26016-99-9
Molecular Formula C3H5Na2O4P
Molecular Weight (g/mol) 182.02
MDL Number MFCD00056853
InChI Key QZIQJIKUVJMTDG-JSTPYPERSA-L
PubChem CID 16397688
IUPAC Name disodium [(2R,3S)-3-methyloxiran-2-yl]phosphonate
SMILES [Na+].[Na+].C[C@@H]1O[C@@H]1P([O-])([O-])=O

Specifications

Type Fosfomycin Disodium
Physical Form Powder
Melting Point >300°C
Color White
Quantity 1 g
Beilstein 4604425
Synonym Phosphomycin disodium salt,, (-)-(1R,2S)-(1,2-Epoxypropyl)phosphonic acid, disodium salt
Solubility Information Freely soluble in water or aqueous solution
Formula Weight 182.02
Percent Purity 90%
Chemical Name or Material Fosfomycin disodium
Show More Show Less
EINECSNumber 247-409-2
RTECSNumber SZ7902000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.