Learn More
Alkali Scientific Fosfomycin [Phosphomycin], Disodium Salt, 1 G
SDP
Catalog No. 5000660478
Encompass_Preferred
Fosfomycin Disodium Salt, 1g is a broad-spectrum antibiotic used for laboratory research, especially in antimicrobial resistance studies. It works by inhibiting bacterial cell wall synthesis, leading to the death of susceptible bacteria. Fosfomycin is particularly useful for treating urinary tract infections and is also employed in microbiological testing to evaluate bacterial susceptibility. The 1g vial is suitable for preparing solutions for in vitro testing and is often used in pharmaceutical research, where it serves as a reference drug in antibiotic efficacy studies and resistance profiling for both Gram-positive and Gram-negative bacteria.
Catalog No. 50-006-60478 Supplier Alkali Scientific Supplier No. FOS001CP
Fosfomycin Disodium Salt, 1g is a broad-spectrum antibiotic used for laboratory research, especially in antimicrobial resistance studies. It works by inhibiting bacterial cell wall synthesis, leading to the death of susceptible bacteria. Fosfomycin is particularly useful for treating urinary tract infections and is also employed in microbiological testing to evaluate bacterial susceptibility. The 1g vial is suitable for preparing solutions for in vitro testing and is often used in pharmaceutical research, where it serves as a reference drug in antibiotic efficacy studies and resistance profiling for both Gram-positive and Gram-negative bacteria.