Learn More
Alkali Scientific Fosfomycin [Phosphomycin], Disodium Salt, 5 G
SDP
Catalog No. 5000659888
Encompass_Preferred
Fosfomycin Disodium Salt, 5g is a broad-spectrum antibiotic used in laboratory research to study bacterial resistance and antimicrobial activity. This antibiotic inhibits bacterial cell wall synthesis, making it effective against a wide range of Gram-positive and Gram-negative bacteria. Fosfomycin is particularly used in treating urinary tract infections and is commonly employed in microbiological testing. The 5g vial is ideal for preparing larger volumes of antibiotic solution for in vitro susceptibility testing, antimicrobial resistance studies, and other pharmaceutical research applications focused on bacterial infections and treatment development.
Catalog No. 50-006-59888 Supplier Alkali Scientific Supplier No. FOS002CP
Fosfomycin Disodium Salt, 5g is a broad-spectrum antibiotic used in laboratory research to study bacterial resistance and antimicrobial activity. This antibiotic inhibits bacterial cell wall synthesis, making it effective against a wide range of Gram-positive and Gram-negative bacteria. Fosfomycin is particularly used in treating urinary tract infections and is commonly employed in microbiological testing. The 5g vial is ideal for preparing larger volumes of antibiotic solution for in vitro susceptibility testing, antimicrobial resistance studies, and other pharmaceutical research applications focused on bacterial infections and treatment development.