Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fosfomycin Sodium, G-Biosciences
SCI_ED

Catalog No. S139758
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139758 100 g
S139716 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139758 Supplier G-Biosciences Supplier No. RC1026
Add to Cart
Add to Cart

Fosfomycin sodium is a phosphoenolpyruvate analog antibiotic isolated from Streptomyces fradiae. It targets gram-positive and gram-negative bacteria, especially E. faecalis and E. coli.

Fosfomycin concentrates in the bladder and kidney, decreasing nephrotoxicity and ototoxicity of anti-tumor agents containing platinum.

Chemical Identifiers

Molecular Weight (g/mol) 182.02 g/mol

Specifications

Boiling Point 342.7°C
Quantity 100 g
Percent Purity 98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.