Learn More
Cayman Chemical Fosfomycin Trometamol, Purity: ≥98%, CAS: 78964-85-9, 5 mg
SDP
Catalog No. 502995208
Encompass_Preferred
A broad-spectrum antibiotic active against Gram-negative and Gram-positive bacteria (MIC90s = 4-32 µg/ml); enters cells via the glycerol-3-phosphate and hexose-6-phosphate transporters; inhibits UDP-GlcNAc enolpyruvyl transferase (MurA)
Catalog No. 50-299-5208 Supplier Cayman Chemical Supplier No. 236325 MG
A broad-spectrum antibiotic active against Gram-negative and Gram-positive bacteria (MIC90s = 4-32 µg/ml); enters cells via the glycerol-3-phosphate and hexose-6-phosphate transporters; inhibits UDP-GlcNAc enolpyruvyl transferase (MurA)